siRNA Details
| Gene Symbol: Arhgef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130150.1 | NP_001123622.1 | 6 | 511 | ||
| NM_001130151.1 | NP_001123623.1 | 6 | 511 | |||
| NM_001130152.1 | NP_001123624.1 | 6 | 520 | |||
| NM_001130153.1 | NP_001123625.1 | 6 | 511 | |||
| NM_008488.2 | NP_032514.1 | 6 | 511 | |||
| GenBank mRNA | AK012156.1 | - | 6 | 461 | ||
| AK043027.1 | - | 6 | 475 | |||
| AK051518.1 | - | 6 | 461 | |||
| AK131181.1 | - | 6 | 408 | |||
| AK157056.1 | - | 6 | 447 | |||
| AK172354.1 | - | 6 | 447 | |||
| BC012488.1 | - | 6 | 471 | 3154558 | ||
| CV783191.1 | - | 6 | 443 | |||
| U58203.1 | - | 6 | 441 | |||

Arhgef1
Genomic Map