siRNA Details
| Gene Symbol: Lgals6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010707.2 | NP_034837.1 | 841 | |||
| GenBank mRNA | AF026794.1 | - | 853 | |||
| EF017942.1 | - | 1168 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lgals6| Gene Symbol: Lgals6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010707.2 | NP_034837.1 | 841 | |||
| GenBank mRNA | AF026794.1 | - | 853 | |||
| EF017942.1 | - | 1168 | ||||