siRNA Details
| Gene Symbol: Limk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK141813.1 | - | 13, 14 | 1751 | ||
| AK170166.1 | - | 13, 14 | 2077 | |||
| AK172083.1 | - | 12, 13 | 1510 | |||
| BC063768.1 | - | 13, 14 | 1731 | |||
| BQ828670.1 | - | 1, 2 | 118 | |||
| U15159.1 | - | 13, 14 | 1746 | |||

Limk1
Genomic Map