siRNA Details
| Gene Symbol: HSBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001537.3 | NP_001528.1 | 3 | 313 | ||
| GenBank mRNA | AF068754.1 | - | 3 | 193 | ||
| AK172732.1 | - | 3 | 178 | |||
| AY320409.1 | - | 3 | 198 | |||
| BC007515.2 | - | 3 | 185 | 3010737 | ||
| BX366956.2 | - | 3 | 245 | |||
| BX537440.1 | - | 3 | 200 | |||
| BX648972.1 | - | 3 | 197 | |||

HSBP1
Genomic Map