siRNA Details
Gene Symbol: Gzmm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB015727.1 | - | 3 | 353 | ||
AB015728.1 | - | 4 | 774 | |||
BC139217.1 | - | 4 | 643 | 8862239 | ||
BC139218.1 | - | 4 | 643 | 8862240 | ||
BC145578.1 | - | 4 | 640 | 9054144 | ||
BC145579.1 | - | 3 | 483 | |||
BE859680.1 | - | 3 | 285 | |||
BG082217.2 | - | 4 | 406 | |||
L76741.1 | - | 4 | 383 |

More Information
Other Species Targeted

Panther Classification:
