siRNA Details
| Gene Symbol: HSBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001537.3 | NP_001528.1 | 3 | 299 | ||
| GenBank mRNA | AF068754.1 | - | 3 | 179 | ||
| AK172732.1 | - | 3 | 164 | |||
| AY320409.1 | - | 3 | 184 | |||
| BC007515.2 | - | 3 | 171 | 3010737 | ||
| BX366956.2 | - | 3 | 231 | |||
| BX537440.1 | - | 3 | 186 | |||
| BX648972.1 | - | 3 | 183 | |||

HSBP1
Genomic Map