siRNA Details
| Gene Symbol: HSBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001537.3 | NP_001528.1 | 3 | 331 | ||
| GenBank mRNA | AF068754.1 | - | 3 | 211 | ||
| AK172732.1 | - | 3 | 196 | |||
| AY320409.1 | - | 3 | 216 | |||
| BC007515.2 | - | 3 | 203 | 3010737 | ||
| BX366956.2 | - | 3 | 263 | |||
| BX537440.1 | - | 3 | 218 | |||
| BX648972.1 | - | 3 | 215 | |||

HSBP1
Genomic Map