siRNA Details
| Gene Symbol: Lta4h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008517.2 | NP_032543.2 | 16 | 1576 | ||
| GenBank mRNA | AK134931.1 | - | 16 | 1562 | ||
| AK182894.1 | - | 2 | 77 | |||
| AK184826.1 | - | 2 | 77 | |||
| AK186946.1 | - | 2 | 77 | |||
| AK219839.1 | - | 5 | 347 | |||
| BC021417.1 | - | 16 | 1570 | 5004113 | ||
| M63848.1 | - | 1542 | ||||

Lta4h
Genomic Map