siRNA Details
| Gene Symbol: Ltbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013589.3 | NP_038617.3 | 24, 25 | 3871 | ||
| GenBank mRNA | AF004874.1 | - | 25, 26 | 4050 | ||
| BC119785.1 | - | 24, 25 | 3855 | 40099725 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ltbp2| Gene Symbol: Ltbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013589.3 | NP_038617.3 | 24, 25 | 3871 | ||
| GenBank mRNA | AF004874.1 | - | 25, 26 | 4050 | ||
| BC119785.1 | - | 24, 25 | 3855 | 40099725 | ||