siRNA Details
| Gene Symbol: Ltbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008520.2 | NP_032546.2 | 7, 8 | 2169 | ||
| GenBank mRNA | AK038587.1 | - | 6, 7 | 901 | ||
| AK080869.1 | - | 7, 8 | 2169 | |||
| AK131126.1 | - | 3, 4 | 2708 | |||
| BC127051.1 | - | 6, 7 | 768 | |||
| L40459.1 | - | 6, 7 | 1285 | |||

Ltbp3
Genomic Map