siRNA Details
| Gene Symbol: Ltk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008523.2 | NP_032549.2 | 13 | 1599 | ||
| NM_203345.2 | NP_976220.2 | 19 | 2445 | |||
| NM_206941.1 | NP_996824.1 | 14 | 1782 | |||
| NM_206942.2 | NP_996825.2 | 18 | 2262 | |||
| GenBank mRNA | AK157478.1 | - | 11 | 1596 | ||
| BC047138.1 | - | 13 | 1599 | 6389834 | ||
| BC055898.1 | - | 10 | 1668 | |||
| BC141299.1 | - | 18 | 2238 | 9055911 | ||
| M90470.1 | - | 19 | 2445 | |||
| X07984.1 | - | 13 | 1573 | |||
| X52621.1 | - | 13 | 1588 | |||

Ltk
Genomic Map