siRNA Details
Gene Symbol: Ly6d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010742.1 | NP_034872.1 | 2 | 147 | ||
GenBank mRNA | BC025135.1 | - | 2 | 142 | 4914126 | |
X63782.1 | - | 2 | 147 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ly6d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010742.1 | NP_034872.1 | 2 | 147 | ||
GenBank mRNA | BC025135.1 | - | 2 | 142 | 4914126 | |
X63782.1 | - | 2 | 147 |