siRNA Details
| Gene Symbol: Ly6d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010742.1 | NP_034872.1 | 3 | 195 | ||
| GenBank mRNA | BC025135.1 | - | 3 | 190 | 4914126 | |
| X63782.1 | - | 3 | 195 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ly6d| Gene Symbol: Ly6d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010742.1 | NP_034872.1 | 3 | 195 | ||
| GenBank mRNA | BC025135.1 | - | 3 | 190 | 4914126 | |
| X63782.1 | - | 3 | 195 | |||