siRNA Details
| Gene Symbol: Il1rl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025602.3 | NP_001020773.1 | 3 | 519 | ||
| NM_001294171.1 | NP_001281100.1 | 3 | 352 | |||
| NM_010743.3 | NP_034873.2 | 3 | 519 | |||
| GenBank mRNA | AK033417.1 | - | 3 | 372 | ||
| AK145118.1 | - | 3 | 373 | |||
| BC051445.1 | - | 3 | 329 | 30073421 | ||
| BY228016.1 | - | 3 | 354 | |||
| D13695.1 | - | 2 | 211 | |||
| M24843.1 | - | 3 | 360 | |||
| Y07519.1 | - | 3 | 363 | |||

Il1rl1
Genomic Map