siRNA Details
| Gene Symbol: Il1rl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025602.3 | NP_001020773.1 | 8 | 1210 | ||
| NM_001294171.1 | NP_001281100.1 | 8 | 1043 | |||
| NM_010743.3 | NP_034873.2 | 8 | 1210 | |||
| GenBank mRNA | AK033417.1 | - | 8 | 1063 | ||
| AK145118.1 | - | 8 | 1064 | |||
| BC051445.1 | - | 8 | 1020 | 30073421 | ||
| D13695.1 | - | 7 | 902 | |||
| M24843.1 | - | 8 | 1051 | |||
| Y07519.1 | - | 8 | 1054 | |||

Il1rl1
Genomic Map