siRNA Details
| Gene Symbol: Ly86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010745.2 | NP_034875.1 | 3 | 330 | ||
| GenBank mRNA | AB007599.1 | - | 3 | 330 | ||
| AK027929.1 | - | 3 | 319 | |||
| BC065783.1 | - | 3 | 235 | |||
| BC096414.1 | - | 3 | 298 | 849991 | ||

Ly86
Genomic Map