siRNA Details
Gene Symbol: Lyz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013590.4 | NP_038618.1 | 2 | 314 | ||
GenBank mRNA | BC061129.1 | - | 2 | 314 | 30296274 | |
X51547.1 | - | 2 | 284 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lyz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013590.4 | NP_038618.1 | 2 | 314 | ||
GenBank mRNA | BC061129.1 | - | 2 | 314 | 30296274 | |
X51547.1 | - | 2 | 284 |