siRNA Details
| Gene Symbol: Tm4sf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008536.3 | NP_032562.1 | 3 | 753 | ||
| GenBank mRNA | AK077326.1 | - | 1 | 256 | ||
| AK133556.1 | - | 254 | ||||
| AK134603.1 | - | 2 | 689 | |||
| AK164718.1 | - | 1 | 257 | |||
| BC083073.1 | - | 3 | 753 | 6337376 | ||
| KC508089.1 | - | 1 | 228 | |||
| L15429.1 | - | 1 | 243 | |||
| L15443.1 | - | 1 | 243 | |||

Tm4sf1
Genomic Map