siRNA Details
| Gene Symbol: Mab21l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010750.3 | NP_034880.1 | 1 | 923 | ||
| GenBank mRNA | AF040945.1 | - | 1 | 910 | ||
| AF228913.1 | - | 1 | 910 | |||
| AK032114.1 | - | 1 | 894 | |||
| AK043552.1 | - | 1 | 923 | |||
| BC014750.1 | - | 1 | 872 | 4526962 | ||

Mab21l1
Genomic Map