siRNA Details
| Gene Symbol: Amacr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008537.4 | NP_032563.2 | 5 | 1108 | ||
| GenBank mRNA | AK002401.1 | - | 5 | 1076 | ||
| AK048249.1 | - | 5 | 1109 | |||
| AK160586.1 | - | 5 | 1111 | |||
| BC015825.1 | - | 5 | 1036 | 3980103 | ||
| U89906.1 | - | 5 | 1016 | |||

Amacr
Genomic Map