siRNA Details
| Gene Symbol: Marcks | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008538.2 | NP_032564.1 | 2 | 1903 | ||
| GenBank mRNA | AK082623.1 | - | 1 | 54 | ||
| AK136452.1 | - | 1 | 72 | |||
| AK151646.1 | - | 1 | 339 | |||
| AK153217.1 | - | 1 | 335 | |||
| BC046601.1 | - | 2 | 1864 | 6311439 | ||
| M60474.1 | - | 2 | 1893 | |||
| X59374.1 | - | 2 | 1794 | |||

Marcks
Genomic Map