siRNA Details
| Gene Symbol: Mxd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016662.4 | NP_057871.2 | 5 | 431 | ||
| GenBank mRNA | AK086190.1 | - | 3 | 1187 | ||
| AK138742.1 | - | 5 | 432 | |||
| BC051970.1 | - | 5 | 434 | 5714980 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mxd3| Gene Symbol: Mxd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016662.4 | NP_057871.2 | 5 | 431 | ||
| GenBank mRNA | AK086190.1 | - | 3 | 1187 | ||
| AK138742.1 | - | 5 | 432 | |||
| BC051970.1 | - | 5 | 434 | 5714980 | ||