siRNA Details
| Gene Symbol: Smad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252481.1 | NP_001239410.1 | 9 | 1494 | ||
| NM_010754.5 | NP_034884.2 | 9 | 1383 | |||
| GenBank mRNA | AF005743.1 | - | 8 | 1111 | ||
| AK007817.1 | - | 9 | 1160 | |||
| AY334552.1 | - | 7 | 962 | |||
| BC021342.1 | - | 9 | 1182 | 5066237 | ||
| BC089184.1 | - | 9 | 1494 | 5322611 | ||
| U60530.1 | - | 8 | 1111 | |||

Smad2
Genomic Map