siRNA Details
| Gene Symbol: Mageb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010759.1 | NP_034889.1 | 1 | 69 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mageb1| Gene Symbol: Mageb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010759.1 | NP_034889.1 | 1 | 69 | ||