siRNA Details
| Gene Symbol: Ccndbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010761.2 | NP_034891.2 | 2 | 232 | ||
| GenBank mRNA | AK144883.1 | - | 2 | 209 | ||
| AK154285.1 | - | 2 | 186 | |||
| AK160205.1 | - | 2 | 198 | |||
| AK163150.1 | - | 2 | 145 | |||
| AK167345.1 | - | 2 | 197 | |||
| BC001984.1 | - | 2 | 168 | 3489973 | ||
| BC005802.1 | - | 1 | 241 | 3707919 | ||
| CO810054.1 | - | 2 | 239 | |||
| U50734.1 | - | 2 | 150 | |||

Ccndbp1
Genomic Map