siRNA Details
| Gene Symbol: Ccndbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010761.2 | NP_034891.2 | 7 | 619 | ||
| GenBank mRNA | AK144883.1 | - | 7 | 596 | ||
| AK154285.1 | - | 7 | 573 | |||
| AK160205.1 | - | 7 | 585 | |||
| AK163150.1 | - | 7 | 532 | |||
| AK167345.1 | - | 7 | 584 | |||
| BC001984.1 | - | 7 | 555 | 3489973 | ||
| BC004673.1 | - | 1 | 39 | |||
| BC005802.1 | - | 6 | 628 | 3707919 | ||
| U50734.1 | - | 7 | 537 | |||

Ccndbp1
Genomic Map