siRNA Details
| Gene Symbol: Mak | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145802.1 | NP_001139274.1 | 8 | 1129 | ||
| NM_001145803.1 | NP_001139275.1 | 9 | 1222 | |||
| NM_008547.2 | NP_032573.2 | 8 | 1117 | |||
| GenBank mRNA | BC050009.1 | - | 9 | 1161 | ||
| X66983.1 | - | 8 | 1111 | |||

Mak
Genomic Map