siRNA Details
| Gene Symbol: Mapkapk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010765.2 | NP_034895.1 | 4, 5 | 986 | ||
| GenBank mRNA | AF039840.1 | - | 4, 5 | 1000 | ||
| AY533679.1 | - | 4, 5 | 294 | |||
| AY533680.2 | - | 4, 5 | 294 | |||
| BC019184.1 | - | 4, 5 | 408 | 5010305 | ||

Mapkapk5
Genomic Map