siRNA Details
| Gene Symbol: Marco | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010766.2 | NP_034896.1 | 16 | 1612 | ||
| GenBank mRNA | AK156388.1 | - | 16 | 1286 | ||
| AK159094.1 | - | 3 | 117 | |||
| AK159967.1 | - | 16 | 1294 | |||
| AK169230.1 | - | 16 | 1288 | |||
| BC132338.1 | - | 16 | 1552 | 40130615 | ||
| BC137939.1 | - | 16 | 1552 | 8860958 | ||
| U18424.1 | - | 16 | 1464 | |||

Marco
Genomic Map