siRNA Details
| Gene Symbol: Mbd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF072248.1 | - | 3 | 581 | ||
| BC038264.1 | - | 3 | 382 | 5250237 | ||
| BQ042794.1 | - | 2 | 149 | |||
| CK625083.1 | - | 3 | 401 | |||
| CX208033.1 | - | 3 | 473 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mbd3| Gene Symbol: Mbd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF072248.1 | - | 3 | 581 | ||
| BC038264.1 | - | 3 | 382 | 5250237 | ||
| BQ042794.1 | - | 2 | 149 | |||
| CK625083.1 | - | 3 | 401 | |||
| CX208033.1 | - | 3 | 473 | |||