siRNA Details
| Gene Symbol: Mbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010775.2 | NP_034905.1 | 5 | 678 | ||
| GenBank mRNA | AK034788.1 | - | 5 | 678 | ||
| BC012245.1 | - | 5 | 640 | 4193682 | ||
| BC021762.1 | - | 5 | 632 | 5132514 | ||
| D11441.1 | - | 5 | 622 | |||
| S42292.1 | - | 5 | 678 | |||

Mbl1
Genomic Map