siRNA Details
Gene Symbol: Mc4r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016977.4 | NP_058673.2 | 1 | 757 | ||
GenBank mRNA | AK136793.1 | - | 1 | 743 | ||
BC116957.1 | - | 1 | 375 | 40126276 | ||
BC116959.1 | - | 1 | 375 | 40126278 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
