siRNA Details
| Gene Symbol: Mcf2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159485.1 | NP_001152957.1 | 5, 6 | 672 | ||
| NM_001159486.1 | NP_001152958.1 | 5, 6 | 672 | |||
| NM_178076.3 | NP_835177.2 | 5, 6 | 575 | |||
| GenBank mRNA | AK048898.1 | - | 5, 6 | 539 | ||
| AK147252.1 | - | 2, 3 | 779 | |||
| AK170186.1 | - | 5, 6 | 633 | |||
| AK170558.1 | - | 5, 6 | 606 | |||
| BC058622.1 | - | 5, 6 | 510 | 5704822 | ||
| DQ978380.1 | - | 4, 5 | 426 | |||
| S76838.1 | - | 6, 7 | 869 | |||

Mcf2l
Genomic Map