siRNA Details
| Gene Symbol: Mcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008562.3 | NP_032588.1 | 2 | 789 | ||
| GenBank mRNA | AK159504.1 | - | 2 | 701 | ||
| AK215970.1 | - | 1 | 14 | |||
| BC003839.1 | - | 2 | 718 | 3491015 | ||
| BC005427.1 | - | 2 | 711 | 3493739 | ||
| BC021638.1 | - | 2 | 728 | 4190996 | ||
| GU182318.1 | - | 3 | 515 | |||
| U35623.1 | - | 2 | 701 | |||

Mcl1
Genomic Map