siRNA Details
| Gene Symbol: HSPA1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC018740.1 | - | 1 | 2209 | 4906865 | |
| Gene Symbol: HSPA1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005346.4 | NP_005337.2 | 1 | 2318 | ||
| GenBank mRNA | AK303967.1 | - | 2 | 2043 | ||
| AL360193.1 | - | 1 | 441 | |||
| BC009322.2 | - | 1 | 2142 | 4123837 | ||
| BC057397.1 | - | 1 | 2299 | 6188453 | ||
| BC063507.1 | - | 1 | 2305 | 4153196 | ||

HSPA1A
Genomic Map