siRNA Details
| Gene Symbol: Mcpt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008571.1 | NP_032597.1 | 3 | 345 | ||
| GenBank mRNA | AK133521.1 | - | 3 | 346 | ||
| AY007570.1 | - | 3 | 345 | |||
| BC119248.1 | - | 3 | 338 | 8733997 | ||
| BC119250.1 | - | 3 | 338 | 8733999 | ||

Mcpt2
Genomic Map