siRNA Details
| Gene Symbol: HSPA1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005527.3 | NP_005518.3 | 2 | 559 | ||
| GenBank mRNA | AK223386.1 | - | 2 | 533 | ||
| AK295416.1 | - | 2 | 793 | |||
| AK302179.1 | - | 2 | 528 | |||
| AK315227.1 | - | 2 | 469 | |||
| BC034483.1 | - | 2 | 536 | 4824979 | ||
| BG717447.1 | - | 2 | 572 | |||
| D85730.1 | - | 2 | 511 | |||

HSPA1L
Genomic Map