siRNA Details
Gene Symbol: Mcpt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010779.2 | NP_034909.2 | 3 | 310 | ||
GenBank mRNA | AB051900.1 | - | 3 | 258 | ||
AK144385.1 | - | 3 | 311 | |||
AY007569.1 | - | 3 | 310 | |||
BC026198.1 | - | 3 | 286 | 1314753 | ||
CJ046459.1 | - | 3 | 313 | |||
M55617.1 | - | 310 | ||||
X68804.1 | - | 3 | 257 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
