siRNA Details
| Gene Symbol: Cma1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010780.2 | NP_034910.1 | 3 | 463 | ||
| GenBank mRNA | BC139454.1 | - | 3 | 452 | 8862476 | |
| BC139457.1 | - | 3 | 452 | 8862479 | ||
| BC139759.1 | - | 3 | 407 | 40055147 | ||
| M68898.1 | - | 3 | 339 | |||
| M73760.1 | - | 1 | 2165 | |||
| X68805.1 | - | 3 | 464 | |||

Cma1
Genomic Map