siRNA Details
| Gene Symbol: Tpsab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031187.4 | NP_112464.4 | 4 | 725 | ||
| GenBank mRNA | AK040500.1 | - | 2 | 1391 | ||
| BC011328.1 | - | 4 | 690 | 3994444 | ||
| L00653.1 | - | 4 | 556 | |||
| U42405.1 | - | 4 | 444 | |||

Tpsab1
Genomic Map