siRNA Details
| Gene Symbol: Mdk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012335.2 | NP_001012335.1 | 4 | 522 | ||
| NM_001012336.2 | NP_001012336.1 | 4 | 519 | |||
| NM_001291481.1 | NP_001278410.1 | 4 | 429 | |||
| NM_001291482.1 | NP_001278411.1 | 3 | 442 | |||
| NM_010784.5 | NP_034914.1 | 4 | 721 | |||
| GenBank mRNA | AK032742.1 | - | 4 | 425 | ||
| AK084457.1 | - | 4 | 442 | |||
| AY864926.1 | - | 4 | 455 | |||
| BC012244.1 | - | 4 | 390 | 4167496 | ||
| CF728526.1 | - | 4 | 415 | |||
| CF731708.1 | - | 4 | 518 | |||
| CF738400.1 | - | 4 | 527 | |||
| CF740555.1 | - | 2 | 239 | |||
| CJ174578.1 | - | 3 | 317 | |||
| M19662.1 | - | 3 | 600 | |||
| M34327.1 | - | 4 | 395 | |||
| M34328.1 | - | 4 | 706 | |||
| M35833.1 | - | 4 | 395 | |||

Mdk
Genomic Map