siRNA Details
| Gene Symbol: Mdk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012335.2 | NP_001012335.1 | 5 | 818 | ||
| NM_001012336.2 | NP_001012336.1 | 5 | 815 | |||
| NM_001291481.1 | NP_001278410.1 | 5 | 725 | |||
| NM_001291482.1 | NP_001278411.1 | 4 | 738 | |||
| NM_001291483.1 | NP_001278412.1 | 4 | 855 | |||
| NM_010784.5 | NP_034914.1 | 5 | 1017 | |||
| GenBank mRNA | AK032742.1 | - | 5 | 721 | ||
| AK084457.1 | - | 5 | 738 | |||
| AY864926.1 | - | 5 | 751 | |||
| CF728526.1 | - | 5 | 711 | |||
| CF740555.1 | - | 3 | 535 | |||
| DQ323886.1 | - | 5 | 793 | |||
| DQ323887.1 | - | 4 | 826 | |||
| M19662.1 | - | 4 | 896 | |||
| M34327.1 | - | 5 | 691 | |||
| M34328.1 | - | 5 | 1002 | |||
| M35833.1 | - | 5 | 691 | |||

Mdk
Genomic Map