siRNA Details
Gene Symbol: Mecom | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021442.2 | NP_067417.1 | 2 | 257 | ||
GenBank mRNA | AJ010015.1 | - | 2 | 257 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mecom | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021442.2 | NP_067417.1 | 2 | 257 | ||
GenBank mRNA | AJ010015.1 | - | 2 | 257 |