siRNA Details
| Gene Symbol: Mef2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170537.1 | NP_001164008.1 | 4 | 762 | ||
| NM_025282.3 | NP_079558.1 | 4 | 762 | |||
| GenBank mRNA | AK009139.1 | - | 4 | 602 | ||
| AK047994.1 | - | 3 | 485 | |||
| AK077603.1 | - | 3 | 551 | |||
| AK132573.1 | - | 4 | 591 | |||
| BC026841.1 | - | 3 | 494 | 4481227 | ||
| BC037731.1 | - | 3 | 490 | 4500786 | ||
| BC057650.1 | - | 3 | 479 | 4500786 | ||
| L13171.1 | - | 2 | 323 | |||

Mef2c
Genomic Map