siRNA Details
| Gene Symbol: Mef2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170537.1 | NP_001164008.1 | 6 | 1117 | ||
| NM_025282.3 | NP_079558.1 | 6 | 1111 | |||
| GenBank mRNA | AK009139.1 | - | 6 | 951 | ||
| AK047994.1 | - | 4 | 696 | |||
| AK077603.1 | - | 5 | 907 | |||
| AK132573.1 | - | 6 | 946 | |||
| BC026841.1 | - | 5 | 849 | 4481227 | ||
| BC037731.1 | - | 5 | 839 | 4500786 | ||
| BC057650.1 | - | 5 | 828 | 4500786 | ||
| L13171.1 | - | 4 | 678 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_008760642, XM_006231736, XM_006231738, XM_006231731, XM_006231739, and others.

Mef2c
Genomic Map