siRNA Details
| Gene Symbol: Met | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008591.2 | NP_032617.2 | 5 | 1858 | ||
| GenBank mRNA | AK148258.1 | - | 5 | 1711 | ||
| BC117969.1 | - | 3 | 1502 | |||
| BC117970.1 | - | 3 | 1498 | |||
| Y00671.1 | - | 3 | 1503 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Met| Gene Symbol: Met | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008591.2 | NP_032617.2 | 5 | 1858 | ||
| GenBank mRNA | AK148258.1 | - | 5 | 1711 | ||
| BC117969.1 | - | 3 | 1502 | |||
| BC117970.1 | - | 3 | 1498 | |||
| Y00671.1 | - | 3 | 1503 | |||