siRNA Details
| Gene Symbol: Mfng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008595.2 | NP_032621.1 | 4 | 661 | ||
| GenBank mRNA | AF015769.1 | - | 4 | 488 | ||
| AK143523.1 | - | 4 | 695 | |||
| BC010983.1 | - | 4 | 683 | 3980692 | ||
| U94349.1 | - | 4 | 661 | |||

Mfng
Genomic Map