siRNA Details
| Gene Symbol: HSPA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002155.4 | NP_002146.2 | 1 | 1843 | ||
| GenBank mRNA | AK093925.1 | - | 1 | 1843 | ||
| AK295212.1 | - | 2 | 1766 | |||
| AK312756.1 | - | 1 | 1843 | |||
| BC004279.1 | - | 1 | 230 | |||
| BC035665.1 | - | 1 | 1833 | 5723718 | ||
| X51758.1 | - | 1 | 662 | |||

HSPA6
Genomic Map