siRNA Details
| Gene Symbol: Clec10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204252.1 | NP_001191181.1 | 9 | 1070 | ||
| NM_010796.3 | NP_034926.1 | 9 | 1067 | |||
| GenBank mRNA | AK140762.1 | - | 8 | 2040 | ||
| AK170779.1 | - | 9 | 1027 | |||
| BC014811.1 | - | 9 | 900 | 4456238 | ||
| S36676.1 | - | 9 | 916 | |||

Clec10a
Genomic Map