siRNA Details
| Gene Symbol: Mgmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008598.2 | NP_032624.1 | 4 | 501 | ||
| GenBank mRNA | AB092489.1 | - | 3 | 412 | ||
| AB092490.1 | - | 3 | 412 | |||
| AK090389.1 | - | 491 | ||||
| BC031888.1 | - | 4 | 470 | 5372377 | ||
| BC096557.1 | - | 4 | 504 | 1366095 | ||
| M84524.1 | - | 3 | 427 | |||

Mgmt
Genomic Map